Learn More
Restek™ Ultra HPLC Columns with C18 Column Packing, 5um Particle Size DFS Item
Similar in pore size to Pinnacle II™ columns, but with greater retention
Supplier:  Restek™ RK9174567
 View more versions of this product
Catalog No. 06-722-236
Only null left
Explore available promotions
Description
Description
  • High-purity silica contains only trace amounts of metal impurities, providing excellent selectivity without the need for mobile phase modifiers
  • With large surface area and high-density bondings
  • 2.5 to 7.5 pH range
  • Temperature limit: 80°C (176°F)
  • C18 columns are good for carbonyls, fatty acids, glycerides, anilines, fat-soluble vitamins, phthalates, barbiturates, steroids, PTH, amino acids, and other acids
Specifications
Specifications
Semi-Preparative Column
150 mm
Spherical
2.5 to 8
80°C
C18
20%
Yes
Column
10 mm
Monomeric C18
5 μm
100 Å
L1
Silica
Reversed Phase
Reversed Phase
Product Suggestions
Product Suggestions