Learn More
Restek™ Ultra HPLC Columns with C18 Column Packing, 5um Particle Size DFS Item
Similar in pore size to Pinnacle II™ columns, but with greater retention
Supplier:  Restek™ RK9174525
 View more versions of this product
Catalog No. 06716326
Only null left
Explore available promotions
Description
Description
  • High-purity silica contains only trace amounts of metal impurities, providing excellent selectivity without the need for mobile phase modifiers
  • With large surface area and high-density bondings
  • 2.5 to 7.5 pH range
  • Temperature limit: 80°C (176°F)
  • C18 columns are good for carbonyls, fatty acids, glycerides, anilines, fat-soluble vitamins, phthalates, barbiturates, steroids, PTH, amino acids, and other acids
Specifications
Specifications
Analytical Column
5 μm
Anilines, Barbiturates, Carbonyls, Fat Soluble Vitamins, Fatty Acids, Glycerides, Phthalates, PTH Amino Acids, Steroids, Other Acids
Monomeric C18
Spherical
2.5 to 8
10 g
L1
Type B Silica
Reversed Phase
Reversed Phase
Column
4.6 mm
1 mL/min
200 mm
4000 psi (275 bar)
5 μm
100 Å
80°C
C18
20%
Yes
Stainless Steel
Product Suggestions
Product Suggestions