Learn More
Restek™ Sulfinert Sample Loops
Ideal for samples containing low concentrations of sulfur or other active compounds. Sulfinert treatment of Restek Gas Sample Loops eliminate active sites in the valve or loop for better recovery of active compounds.
Supplier:  Restek™ RK 22849
Encompass
 View more versions of this product
Catalog No. 06712066
Only null left
Description
Description
  • Sample loop sizes from 5μL to 5mL
  • 0.0625 in. fittings
  • 1mm I.D.
  • For “W Type” Valves
Specifications
Chemical Identifiers
Sulfinert treated
Gas Sample Loop
Specifications
Sulfinert treated
2 mL
Gas Sample Loop
Product Suggestions
Product Suggestions