Learn More
Restek™ Sulfinert Sample Loops
Ideal for samples containing low concentrations of sulfur or other active compounds. Sulfinert treatment of Restek Gas Sample Loops eliminate active sites in the valve or loop for better recovery of active compounds.
Supplier: Restek™ RK 22841
Description
- Sample loop sizes from 5μL to 5mL
- 0.0625 in. fittings
- 1mm I.D.
- For “W Type” Valves
Chemical Identifiers
| Sulfinert treated | |
| Gas Sample Loop |
Specifications
| Sulfinert treated | |
| 10 μL | |
| Gas Sample Loop |