missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cyclosplitter

For use with dirty samples

$648.30

Specifications

Length (Metric) 78.5 mm
Diameter (Metric) Inner 4.0 mm
Diameter (Metric) Outer 6.3 mm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
View More Specs

Products 1
Catalog Number Mfr. No. Liner Material Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Liner Material Product Type Quantity Price Quantity & Availability  
06717796
Encompass
View Documents Promotion Details
Restek™
RK RK20707
Borosilicate Glass IP CycloSplitter Inlet Liner 5/Pk.
Pack of 5 Each for $648.30
Only null left
 
Description

Description

  • Incorporate a cylindrical glass screw in the sample pathway to mix sample and trap nonvolatile residues
  • 4mm I.D.
  • Perform multiple injections before cleaning is required

Specifications

Specifications

78.5 mm
4.0 mm
6.3 mm
Agilent GCs Equipped with Split/Splitless Inlets
Videos
SDS
Documents

Documents