Learn More
Restek™ Rtx™-5MS Capillary Columns DFS Item
Developed specifically for GC/MS.
Supplier:  Restek™ RK12654
 View more versions of this product
Catalog No. 06709872
Only null left
Explore available promotions
Description
Description
  • Nonpolar; 5% diphenyl/95% dimethyl polysiloxane
  • Exceptionally low bleed at high temperatures
  • Undergo rigorous QA testing to ensure they exceed the requirements of even the most sensitive MS
Pesticides, PCBs, Aromatic hydrocarbons, Environmental samples, Essential oils
Specifications
Specifications
0.32 mm
Rtx™-5 MS
Essential Oils Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
-60°C to +325/350°C
30 m
Low polar
G27, G36
350°C
1 μm
1 μm
Stainless Steel
Fused Silica
Product Suggestions
Product Suggestions