missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Rtx™ -5 Capillary Columns - 10m Length

Catalog No. 06713164
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
0.10 mm
0.18 mm
Film Thickness:
0.10 μm
0.20 μm
Particle Size:
0.1 μm
0.2 μm
Temperature Range (Metric):
-60°C to +325/340°C
-60°C to +330/350°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Film Thickness Particle Size Temperature Range (Metric)
06713164 0.18 mm 0.20 μm 0.2 μm -60°C to +325/340°C
06713216 0.10 mm 0.10 μm 0.1 μm -60°C to +330/350°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06713164 Supplier Restek™ Supplier No. RK 40201

General-purpose, fused silica column for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.

All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Low-polarity phase
  • Crossbond diphenyl dimethyl polysiloxane
  • Equivalent to USP G27 and G36 phases

Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.

Order Info

Custom lengths and film thicknesses available.

Specifications

For Use With (Application) Hydrocarbon Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
Particle Size 0.2 μm
Film Thickness 0.20 μm
Length (Metric) 10 m
Material Stainless Steel
Polarity Low polar
Coating Fused Silica
USP Type G27, G36
Stationary Phase Rtx™-5
Type Rtx-5 (G27) Column
Diameter (Metric) Inner 0.18 mm
Max. Temperature (Metric) 350°C
Temperature Range (Metric) -60°C to +325/340°C
Show More Show Less