Learn More
Restek™ Rtx™-1 Capillary Columns DFS Item
Similar in polarity to DB-1 and HP-1
Supplier:  Restek™ RK10139
 Afficher plus de versions de ce produit
Numéro de catalogue. 06708322
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Nonpolar; coated with crossbonded polydimethylsiloxane (100% dimethylpolysiloxane)
  • Manufactured from high-quality fused silica tubing
  • Surrounded by a rugged stainless-steel cage
  • Excellent thermal stability for low bleed at high temperatures
Solvents, Petroleum products, Pharmaceuticals, Waxes, Fuel oils
Spécifications
Spécifications
0.32 mm
Rtx™-1
Drugs of Abuse Analysis, PCB Congener Analysis, Oxygenates Analysis, PCB Congener Analysis, Simulated Distillation, Natural Gas Odorants Analysis, Sulfur Compounds Analysis, Natural Gas Odorants Analysis, Hydrocarbons, Essential Oils, Hydrocarbons, Semivolatiles Analysis
-60°C to +330/350°C
30 m
Non polar
1/Pk.
G1, G2, G38
350°C
0.5 μm
0.5 μm
Stainless Steel
Fused Silica
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS