missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Presents SGE Gas-Tight Syringes Fitted with Removable Needle Valve

Catalog No. 06712540
Encompass
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
Model:
100R-V-GT
1MDR-V-GT
2.5MDR-V-GT
250R-V-GT
500R-V-GT
50R-V-GT
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
06712540 100 μL 100R-V-GT
06712541 250 μL 250R-V-GT
06712542 500 μL 500R-V-GT
06712543 1 mL 1MDR-V-GT
06712544 2.5 mL 2.5MDR-V-GT
06719049 50 μL 50R-V-GT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06712540 Supplier Restek™ Supplier No. RK 24881
Only null left

Ideal for sample storage and transportation.

  • Syringe fitted with removable needle and valve
  • Push-pull shut-off valve

Specifications

Needle Gauge 23 G
Diameter (English) Inner Needle 0.0132 in.
Diameter (Metric) Inner Needle 0.34 mm
Diameter (English) Outer Needle 0.025 in.
Diameter (Metric) Outer Needle 0.64 mm
For Use With (Application) For sample storage and transportation.
Includes Shut-off valves
Product Type SGE Gas-Tight Syringe Fitted with Removable Needle Valve
Model 100R-V-GT
Quantity Ea.
Volume (Metric) 100 μL
Connection Type Removable
Show More Show Less