Learn More
Restek™ Presents SGE Gas-Tight Syringes Fitted with Luer-lock Valve
Catalog No. 06712548
Encompass
Change view
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Model:
100MR-VLLMA-GT
10MDR-VLLMA-GT
1MR-VLL-GT
2.5MDR-VLL-GT
25MDR-VLLMA-GT
50MR-VLLMA-GT
5MDR-VLLMA-GT
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
06712548 10 mL 10MDR-VLLMA-GT
06712545 1 mL 1MR-VLL-GT
06712547 5 mL 5MDR-VLLMA-GT
06712546 2.5 mL 2.5MDR-VLL-GT
06712549 25 mL 25MDR-VLLMA-GT
06712550 50 mL 50MR-VLLMA-GT
06712551 100 mL 100MR-VLLMA-GT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 06712548 Supplier Restek™ Supplier No. RK 24889
Ideal for sample storage and transportation.
  • With Push-pull shut-off valve
  • Fitted with Luer Lock valve
Specifications
For Use With (Application) For sample storage and transportation.
Includes Luer-lock valve
Product Type SGE Gas-Tight Syringe Fitted with Luer lock Valve
Model 10MDR-VLLMA-GT
Quantity Ea.
Volume (Metric) 10 mL