missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Presents Hamilton Syringes for Waters U6K Valves

Catalog No. 06711490
Encompass
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
Model:
801
802
805
810
825
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
06711490 10 μL 801
06711491 25 μL 802
06711492 50 μL 805
06711493 100 μL 810
06711494 250 μL 825
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06711490 Supplier Restek™ Supplier No. RK 21255

For Waters U6K Valves.

  • Reinforced plungers
  • Removable needles
  • Replaceable plunger/barrel assemblies
  • Barrel stop prevents plunger blowout
  • Needle Gauge: 25s
  • Needle Length: 1.97 in.
  • Needle Point Style: 3

Specifications

Needle Gauge 25s G
Length (English) Needle 1.97 in.
Product Type Hamilton Syringes for Waters U6K Valves
Model 801
Needle Point Number 3
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style 3
For Use With (Equipment) Waters U6K Valves
Connection Type Removable