missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Presents Hamilton Priming Syringe for Waters 6000, 6000A, 501, 510, 590, 610 and 610E HP

Catalog No. 06711500
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06711500 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06711500 Supplier Restek™ Supplier No. RK 21265

For Waters 6000, 6000A, 501, 510, 590, 610, and 610E HPLC Pumps.

  • Designed for maximum safety with metal flange and luer lock (LL) hub
  • PTFE-tipped plunger
  • Pressure tested to 700psig
  • Volume: 10mL

Specifications

Material PTFE (Plunger), PTFE (Tip)
Product Type Priming Syringe
Max. Pressure 700 psig
Quantity 1/Ea.
Volume (Metric) 10 mL
For Use With (Equipment) Waters 6000, 6000A, 501, 510, 590, 610, and 610E HPLC Pumps
Connection Type Luer Lock