missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Graduated Safety-Coated Bottles

Catalog No. 06712847
Encompass
Change view
Click to view available options
Volume (Metric):
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06712847 1 L
06712848 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06712847 Supplier Restek™ Supplier No. RK 25304

Graduated, safety-coated bottles with GL-45 threads.

Available as 1 L or 2 L bottles

Specifications

Includes Solid cap
Material Glass
Thread Size GL-45
Volume (Metric) 1 L
For Use With (Application) Mobile phase management
Type Graduated Bottle
Color Clear