Learn More
Restek™ Mininert Precision Sampling Valves for Vials
Catalog No. 06719055
Encompass
Change view
Click to view available options
For Use With:
Crimp-Top Vial
Screw-Thread Vials
Thread Size:
13 mm/425
15 mm/425
18 mm/400
20 mm/400
24 mm/400
20 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With Thread Size
06719055 Crimp-Top Vial 20 mm
06719050 Screw-Thread Vials 13 mm/425
06719052 Screw-Thread Vials 18 mm/400
06719053 Screw-Thread Vials 20 mm/400
06719054 Screw-Thread Vials 24 mm/400
06719051 Screw-Thread Vials 15 mm/425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06719055 Supplier Restek™ Supplier No. RK 24905
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Convenient valves for repetitive sampling and limiting content exposure to the silicon septum.
  • Available for both screw-cap and crimp-top vials
  • Crimp-top valve for 20mm ID glassware slides into the neck of the vial
  • Turn the threaded flange to secure a tight fit
Specifications
Type Valve
For Use With (Application) Precision Sampling
Packaging Quantity 12 Pack
Thread Size 20 mm
For Use With Crimp-Top Vial