Learn More
Restek™ Jumbo Syringes
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
$2,883.49 - $4,729.21
Description
- Adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time
- Central port supplied with a luer lock fitting
- Secondary port is supplied with a septum nut, which enables access to gas sample for adding standards or removing a subsample
- Detachable plunger stem for easy sample storage
Specifications
| Syringe |