Learn More
Restek™ Hamilton Gas-tight Syringes
Catalog No. 06712356
Encompass
Change view
Click to view available options
Volume (Metric):
100 μL
500 μL
2.5 mL
5 mL
10 mL
Model:
1002
1005
1010
1010RN
1710TLL
1750
1710LT
Needle Point Style:
2
Luer
PTFE luer lock
Removable
Removable Bevel
Product Type:
Gastight Syringe
Hamilton PTFE Tip, Gas-Tight Syringes
Hamilton Syringe
Hamilton Syringe with 1010 Removable Needle
Hamilton Syringe with 1710 PTFE Luer lock
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model Needle Point Style Product Type
06712356 100 μL 1710TLL PTFE luer lock Hamilton Syringe with 1710 PTFE Luer lock
06712355 100 μL 1710LT Luer Hamilton Syringe
06712363 500 μL 1750 Luer Gastight Syringe
06712372 2.5 mL 1002 2 Hamilton PTFE Tip, Gas-Tight Syringes
06712376 5 mL 1005 Removable Bevel Gastight Syringe
06712379 10 mL 1010RN Removable Hamilton Syringe with 1010 Removable Needle
06712380 10 mL 1010 Luer Gastight Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Fisher Catalog Numbers: 06-712-355, -356 and -363 do NOT include a needle. The needle can be purchased separately (Fisher Catalog Number: 06718945).
Catalog No. 06712356 Supplier Restek™ Supplier No. RK 24566
Fisher Catalog Numbers: 06-712-355, -356 and -363 do NOT include a needle. The needle can be purchased separately (Fisher Catalog Number: 06718945).
Suitable for gases or liquids, for maximum inertness.
  • High accuracy of dispensed volumes
  • Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
For Use With (Application) Gases or liquids, optimum septum penetration and prevention of septum coring
Product Type Hamilton Syringe with 1710 PTFE Luer lock
Model 1710TLL
Quantity Ea.
Volume (Metric) 100 μL
Needle Point Style PTFE luer lock