missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resorcinol Monoacetate 50.0+%, TCI America™
$76.43 - $443.65
Chemical Identifiers
| CAS | 102-29-4 |
|---|---|
| Molecular Formula | C8H8O3 |
| Molecular Weight (g/mol) | 152.149 |
| MDL Number | MFCD00002266 |
| InChI Key | ZZPKZRHERLGEKA-UHFFFAOYSA-N |
| Synonym | resorcinol monoacetate, acetylresorcinol, euresol, remonol, 3-acetoxyphenol, resorcitate, m-hydroxyphenyl acetate, 1,3-benzenediol, monoacetate, resorcinol, monoacetate, resorcinol acetate |
| PubChem CID | 5055 |
| ChEBI | CHEBI:29672 |
| IUPAC Name | (3-hydroxyphenyl) acetate |
| SMILES | CC(=O)OC1=CC=CC(=C1)O |
Chemical Identifiers
| 102-29-4 | |
| 152.149 | |
| ZZPKZRHERLGEKA-UHFFFAOYSA-N | |
| 5055 | |
| (3-hydroxyphenyl) acetate |
| C8H8O3 | |
| MFCD00002266 | |
| resorcinol monoacetate, acetylresorcinol, euresol, remonol, 3-acetoxyphenol, resorcitate, m-hydroxyphenyl acetate, 1,3-benzenediol, monoacetate, resorcinol, monoacetate, resorcinol acetate | |
| CHEBI:29672 | |
| CC(=O)OC1=CC=CC(=C1)O |
Specifications
| 102-29-4 | |
| C8H8O3 | |
| 25 g | |
| ZZPKZRHERLGEKA-UHFFFAOYSA-N | |
| (3-hydroxyphenyl) acetate | |
| 5055 | |
| 152.15 | |
| Liquid |
| 283°C | |
| MFCD00002266 | |
| resorcinol monoacetate, acetylresorcinol, euresol, remonol, 3-acetoxyphenol, resorcitate, m-hydroxyphenyl acetate, 1,3-benzenediol, monoacetate, resorcinol, monoacetate, resorcinol acetate | |
| CC(=O)OC1=CC=CC(=C1)O | |
| 152.149 | |
| CHEBI:29672 | |
| ≥50.0% (GC) | |
| Resorcinol Monoacetate |
Safety and Handling
EINECSNumber : (3)-2785
RTECSNumber : VH2750000
TSCA : Yes