CAS RN 102-29-4
IUPAC Name:
3-hydroxyphenyl acetate
Synonyms:
acetylresorcinol resorcitate M-hydroxyphenyl acetate euresol remonol
Molecular Weight (g/mol):
152.15
Molecular Formula:
C8H8O3
InChi Key:
ZZPKZRHERLGEKA-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=CC(O)=C1
1
–
1
of
1
results
1
Resorcinol Monoacetate 50.0+%, TCI America™
CAS: 102-29-4 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.149 MDL Number: MFCD00002266 InChI Key: ZZPKZRHERLGEKA-UHFFFAOYSA-N Synonym: resorcinol monoacetate,acetylresorcinol,euresol,remonol,3-acetoxyphenol,resorcitate,m-hydroxyphenyl acetate,1,3-benzenediol, monoacetate,resorcinol, monoacetate,resorcinol acetate PubChem CID: 5055 ChEBI: CHEBI:29672 IUPAC Name: (3-hydroxyphenyl) acetate SMILES: CC(=O)OC1=CC=CC(=C1)O