missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resorcinol Monoacetate 50.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 283°C |
| Quantity | 500 g |
| Formula Weight | 152.15 |
| Percent Purity | ≥50.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Resorcinol Monoacetate |
Safety and Handling
| EINECSNumber | (3)-2785 |
| RTECSNumber | VH2750000 |
| TSCA | Yes |