missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol Monoacetate 50.0+%, TCI America™

Catalog No. R000925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R000925G 25 g
R0009500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R000925G Supplier TCI America Supplier No. R000925G
Only null left

Chemical Identifiers

CAS 102-29-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002266
InChI Key ZZPKZRHERLGEKA-UHFFFAOYSA-N
Synonym resorcinol monoacetate, acetylresorcinol, euresol, remonol, 3-acetoxyphenol, resorcitate, m-hydroxyphenyl acetate, 1,3-benzenediol, monoacetate, resorcinol, monoacetate, resorcinol acetate
PubChem CID 5055
ChEBI CHEBI:29672
IUPAC Name (3-hydroxyphenyl) acetate
SMILES CC(=O)OC1=CC=CC(=C1)O

Specifications

Boiling Point 283°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥50.0% (GC)
Physical Form Liquid
Chemical Name or Material Resorcinol Monoacetate
EINECSNumber (3)-2785
RTECSNumber VH2750000
TSCA Yes