missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, Flake, Technical, Spectrum™ Chemical

Catalog No. p-7166251
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Poly Pail
Catalog No. Quantity Packaging
18613026 500 g Poly Pail
18613024 2.5 kg Poly Pail
18613025 25 kg Poly Pail
3 options

Catalog No. 18613026

Supplier: Spectrum Chemical Mfg Cor R1529500GM

Only null left

Chemical Identifiers

CAS 108-46-3

Specifications

CAS Max % 1
Boiling Point 277 to 281°C
Packaging Poly Pail
Quantity 500 g
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.