missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, 99%

CAS: 108-46-3 | C6H6O2 | 110.112 g/mol

Supplier:  Thermo Scientific Chemicals A130800I

 View more versions of this product

Catalog No. AAA130800I


Only null left
Explore available promotions

Description

Description

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

Solubility
Soluble in water, alcohol, ether and glycerol.

Notes
Stable. May discolor on exposure to air or light.
Specifications

Specifications

Resorcinol
109°C to 112°C
4 to 6
171°C (339°F)
C6H6O2
5000 g
906905
14,8155
Soluble in water,alcohol,ether and glycerol.
C1=CC(=CC(=C1)O)O
110.112
CHEBI:27810
99%
108-46-3
1.272
280°C to 281°C
Phenol-like
MFCD00002269
UN2876
Air and light sensitive
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
5054
110.11

Chemical Identifiers

108-46-3
110.112
GHMLBKRAJCXXBS-UHFFFAOYSA-N
5054
benzene-1,3-diol
C6H6O2
MFCD00002269
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
CHEBI:27810
C1=CC(=CC(=C1)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H301-H315-H319-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: RESORCINOL

EINECSNumber : 203-585-2

RTECSNumber : VG9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only