missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, 99%

Catalog No. AAA1308018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1308018 50 g
AAA1308030 250 g
AAA130800B 1000 g
AAA130800I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1308018 Supplier Thermo Scientific Chemicals Supplier No. A1308018

CAS: 108-46-3 | C6H6O2 | 110.112 g/mol

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

Solubility
Soluble in water, alcohol, ether and glycerol.

Notes
Stable. May discolor on exposure to air or light.

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 109°C to 112°C
Density 1.272
pH 4 to 6
Boiling Point 280°C to 281°C
Flash Point 171°C (339°F)
Odor Phenol-like
Quantity 50 g
UN Number UN2876
Beilstein 906905
Sensitivity Air and light sensitive
Merck Index 14,8155
Solubility Information Soluble in water,alcohol,ether and glycerol.
Formula Weight 110.11
Percent Purity 99%
Chemical Name or Material Resorcinol
Show More Show Less
Hazard Category H301-H315-H319-H500
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: RESORCINOL
EINECSNumber 203-585-2
RTECSNumber VG9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only