missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, 98%

Catalog No. p-190688
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132290500 50 g Plastic bottle
AC132292500 250 g Plastic bottle
AC132290010 1 kg Plastic bottle
AC132290050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC132290500 Supplier Thermo Scientific Chemicals Supplier No. 132290500

CAS: 108-46-3 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 109.0°C to 111.0°C
Color Colorless to Yellow
Boiling Point 281.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 50 g
Beilstein 06,II,802
Merck Index 15,8276
Solubility Information Solubility in water: 140 g/100 mL. Other solubilities: freely soluble in ether, glycerol, soluble in benzene and amyl alcohol, slightly soluble in chloroform
Formula Weight 110.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Resorcinol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-585-2
RTECSNumber VG9625000
TSCA TSCA

RUO – Research Use Only