missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Resiquimod | |
| 193°C | |
| C17H22N4O2 | |
| 100 mg | |
| BXNMTOQRYBFHNZ-UHFFFAOYSA-N | |
| 1-[4-amino-2-(ethoxymethyl)imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol | |
| 159603 | |
| 314.39 | |
| Crystalline Powder |
| 144875-48-9 | |
| 7.2 | |
| MFCD00937759 | |
| resiquimod, 1-4-amino-2-ethoxymethyl-1h-imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol, resiquimod inn, unii-v3dmu7pvxf, v3dmu7pvxf, resiquimod r-848, 4-amino-2-ethoxymethyl-a,a-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, alpha-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, 1-4-amino-2-ethoxymethyl imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol | |
| CCOCC1=NC2=C(N1CC(C)(C)O)C3=CC=CC=C3N=C2N | |
| 314.389 | |
| CHEBI:36706 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 144875-48-9 | |
| 314.389 | |
| BXNMTOQRYBFHNZ-UHFFFAOYSA-N | |
| 159603 | |
| 1-[4-amino-2-(ethoxymethyl)imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol |
| C17H22N4O2 | |
| MFCD00937759 | |
| resiquimod, 1-4-amino-2-ethoxymethyl-1h-imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol, resiquimod inn, unii-v3dmu7pvxf, v3dmu7pvxf, resiquimod r-848, 4-amino-2-ethoxymethyl-a,a-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, alpha-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, 1-4-amino-2-ethoxymethyl imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol | |
| CHEBI:36706 | |
| CCOCC1=NC2=C(N1CC(C)(C)O)C3=CC=CC=C3N=C2N |
Safety and Handling
RTECSNumber : NJ5911320
TSCA : No
Recommended Storage : Freezer (-20 °C)