missing translation for 'onlineSavingsMsg'
Learn More

Resiquimod 98.0+%, TCI America™

Catalog No. p-7135751
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R019725MG 25 mg
R0197100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R019725MG Supplier TCI America Supplier No. R019725MG
Only null left

Chemical Identifiers

CAS 144875-48-9
Molecular Formula C17H22N4O2
Molecular Weight (g/mol) 314.389
MDL Number MFCD00937759
InChI Key BXNMTOQRYBFHNZ-UHFFFAOYSA-N
Synonym resiquimod, 1-4-amino-2-ethoxymethyl-1h-imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol, resiquimod inn, unii-v3dmu7pvxf, v3dmu7pvxf, resiquimod r-848, 4-amino-2-ethoxymethyl-a,a-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, alpha-dimethyl-1h-imidazo 4,5-c quinoline-1-ethanol, 1-4-amino-2-ethoxymethyl imidazo 4,5-c quinolin-1-yl-2-methylpropan-2-ol
PubChem CID 159603
ChEBI CHEBI:36706
IUPAC Name 1-[4-amino-2-(ethoxymethyl)imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol
SMILES CCOCC1=NC2=C(N1CC(C)(C)O)C3=CC=CC=C3N=C2N

Specifications

Melting Point 193°C
pH 7.2
Quantity 25 mg
Formula Weight 314.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Resiquimod
RTECSNumber NJ5911320
TSCA No
Recommended Storage Freezer (-20 °C)