Learn More
MilliporeSigma™ Supelco™ Reservoir for Filtration Apparatus 1
Replacement glass reservoir for Supelco™ mobile phase filtration apparatus 1
Supplier: MilliporeSigma™ Supelco™ 58074
Description
Reservoir for Filtration Apparatus 1 protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.Chemical Identifiers
| Liquid Chromatography | |
| Filtration Apparatus 1 |
Specifications
| Liquid Chromatography | |
| Filtration Apparatus 1 | |
| 500 mL |