missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Research Grade Fetal Bovine Serum, US Sourced
Research Grade Fetal Bovine Serum that is US sourced and triple 0.1 μm filtered.
$116.44
Specifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG - Nephelometer |
|---|---|
| Age | Fetal |
| Country of Origin | United States |
| Form | Liquid |
| Format | Bottle |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
FB12999103
|
Fisherbrand™
SH3A11774.01 |
50 mL |
Each for $116.44
|
|
|||||
Description
- US Origin is compliant with current good manufacturing practices (cGMP).
- For research use only.
Specifications
| Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG - Nephelometer | |
| United States | |
| Bottle | |
| Sterile-filtered | |
| Triple-filtered, 0.1 μm | |
| Yes | |
| No | |
| No |
| Fetal | |
| Liquid | |
| Fetal Bovine Serum | |
| Sterile-filtered | |
| Research | |
| For Further Manufacturing Or Research Use. Not For Diagnostic Or Therapeutic Use. | |
| < 35 mg/dL |