missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Research Grade Fetal Bovine Serum, US Sourced

Catalog No. FB12999103
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB12999103 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB12999103 Supplier Fisherbrand™ Supplier No. SH3A11774.01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Research Grade Fetal Bovine Serum that is US sourced and triple 0.1 μm filtered.

  • US Origin is compliant with current good manufacturing practices (cGMP).
  • For research use only.

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG - Nephelometer
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Sterile-filtered
Sterility Sterile-filtered
Sterilization Method Triple-filtered, 0.1 μm
Grade Research
Processing Yes
For Use With (Application) For Further Manufacturing Or Research Use. Not For Diagnostic Or Therapeutic Use.
Heat Inactivated No
Hemoglobin Concentration < 35 mg/dL
Lyophilized No
Show More Show Less