missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative E. coli DNA Kit with PrepSEQ™ Residual DNA Sample Preparation Kit, 100 reactions

Catalog No. 4460366
Encompass
Change view
Click to view available options
Assay Kit and Sample Preparation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
4460366 Assay Kit and Sample Preparation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4460366 Supplier Applied Biosystems™ Supplier No. 4460366

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ E. coli Residual DNA Quantitation System, enables highly sensitive quantitation of host cell DNA from E. coli expression systems, typically in less than 4 hours.

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ E. coli Residual DNA Quantitation System, enables highly sensitive quantitation of host cell DNA from E. coli expression systems, typically in less than 4 hours. The kit is part of an integrated system that includes sample preparation, TaqMan™ Assay and master mix, standard DNA, instrument, and software. The system overcomes the limitations of traditional methods by combining high-recovery PrepSEQ™ sample preparation and TaqMan™ Assay-based quantitation of residual DNA.

Features of the resDNASEQ Quantitative E. coli DNA Kit include:
• Highly sensitive quantitation using proven TaqMan™ real-time qPCR technology
• Manual and automated sample preparation, optimized for quantitative recovery from complex sample matrices (Table 1)
• Consistent performance across the expected range of DNA fragment sizes
• Integrated system from sample to results with sample preparation, master mix, TaqMan™ primer/probe mix, and genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ™ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Rapid, Sensitive Detection of E.Coli DNA
The resDNASEQ™ E. coli Residual DNA Quantitation System is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from E. coli, an expression system commonly used for the production of recombinant proteins. Reliable and rapid, the system enables sensitive (LOQ = 15 pg DNA/mL test sample) and specific quantitation of E. coli DNA, typically in less than 4 hours. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types—from in-process samples to bulk drug substances—whether a sample contains high molecular weight or sheared DNA.

Table 1. DNA recovery using manual PrepSEQ™ sample preparation protocol.
Assay performance data using 10 pg E. coli genomic DNA spike per sample, 3 analysts, and 9 test samples.

  50 mg/mL protein sample 
  Mean Recovery Mean %CV
E. coli83%5.04%


Specifications

Content And Storage resDNASEQ E.coli DNA Control
• E.coli DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ E.coli DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• E.coli DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell E.coli
Label or Dye FAM
Product Line resDNASEQ™
Quantity 100 reactions
No. of Reactions 100 Reactions
Includes Assay Kit and Sample Preparation

For Research Use Only. Not for use in diagnostic procedures.