missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Human Residual DNA Quantitation Kits
GREENER_CHOICE

Catalog No. A27335
Change view
Click to view available options
Includes:
Assay Kit and Sample Preparation
Assay Kit only
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
A27335 Assay Kit and Sample Preparation
A26366 Assay Kit only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27335 Supplier Applied Biosystems™ Supplier No. A27335
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The resDNASEQ Human Residual DNA Quantitation Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from human cell lines used as expression systems for recombinant proteins.

The resDNASEQ Human Residual DNA Quantitation Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from human cell lines used as expression systems for recombinant proteins. Reliable and rapid, the resDNASEQ system enables sensitive (as low as 15 pg DNA/mL test sample) and specific quantitation of human cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples to final product, whether the sample contains high molecular weight or sheared DNA.

Features of the resDNASEQ Human Residual DNA Quantitation Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, TaqMan primer/probe mix, and human genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

The resDNASEQ Human Residual DNA Quantitation Kit provides highly sensitive detection of human DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan real-time qPCR technology allows you to test samples with variable levels of human DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.

Easy to use
With the resDNASEQ Human Residual DNA Quantitation Kit you perform qPCR analysis to compare the amount of human DNA in your test samples to a standard curve generated with known amounts of purified human genomic DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to human genomic DNA standard, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of residual human DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ system you get consistent assay performance across a range of DNA fragment sizes.

*Due to the high degree of similarity between genomes, DNA from closely-related primate species such as monkey (Vero), chimpanzee, and orangutan are expected be detected by the resDNASEQ Human Quantitation Kit.

Specifications

Content And Storage resDNASEQ Human DNA Control
• Human DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ Human DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• Human DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell Human DNA
Label or Dye FAM
Product Line resDNASEQ
Quantity 100 reactions
No. of Reactions 100 Reactions
Includes Assay Kit and Sample Preparation
I would like to quantify cell-free DNA using a qPCR-based kit. Can I use the resDNASEQ Human Residual DNA Quantitation Kit (Cat. No. A26366)?

We have not tested the resDNASEQ Human Residual DNA Quantitation Kit (Cat. No. A26366) for quantitation of cell free DNA. You would need a cell free DNA isolation kit to extract the cell free DNA. Please take a look at the following link for more information on cell free DNA isolation. Once the cell free DNA is extracted you can use any of the pre-designed or custom TaqMan assays specific for the target gene.