missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ dPCR E. coli DNA Kit
GREENER_CHOICE

Catalog No. A57017
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57017 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A57017 Supplier Applied Biosystems™ Supplier No. A57017
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems resDNASEQ dPCR E. coli DNA Kit is a digital PCR (dPCR)-based solution to detect host-cell DNA from E. coli, an expression system commonly used for production of recombinant proteins and plasmids.

The Applied Biosystems resDNASEQ dPCR E. coli DNA Kit is a digital PCR (dPCR)-based solution to detect host-cell DNA from E. coli, an expression system commonly used for production of recombinant proteins and plasmids. Reliable and rapid, the assay enables sensitive and specific quantification of E. coli DNA, helping provide confidence in quantitation data obtained from a broad range of sample types, from in-process samples with different sample matrices to formulations with final products.

Features of the resDNASEQ dPCR E.coli DNA Kit include:

  • Accurate, absolute quantitation of residual host-cell E. coli DNA
  • Highly sensitive quantitation using proven TaqMan Assays, with results in under three hours
  • Highly characterized E. coli DNA control that can be leveraged during qualification and verification
  • Easy-to-use, integrated sample-to-results kit consisting of Absolute Q DNA Digital PCR Master Mix, E. coli assay mix, well-characterized DNA positive control, and DNA dilution buffer
  • Manual or automated sample preparation, optimized for quantitative recovery of samples from complex matrices

Integrated workflow solution
The resDNASEQ dPCR E. coli DNA Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The kit has been validated on the QuantStudio Absolute Q Digital PCR System. The Absolute Q system has a high tolerance of common inhibitors in biopharmaceutical sample matrices, eliminating the need for nucleic acid extraction in some cases. If nucleic acid extraction is required, we recommend use of the PrepSEQ Residual DNA Sample Preparation Kit, which offers both a manual and automated sample preparation workflow.

Easy to use
With the resDNASEQ dPCR E. coli DNA Kit, dPCR analysis is performed to quantitate the copy numbers of residual E.coli DNA in the test samples. The resDNASEQ kit includes almost everything needed to carry out these steps, from master mix to TaqMan primer/probe mix to E. coli genomic DNA control, as well as step-by-step instructions. A copy-to-mass conversion tool is also provided to guide you on the final result conversion to satisfy regulation requirements.

Specifications

Content And Storage resDNASEQ E.coli DNA Control
• E.coli DNA Control, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ E.coli DNA dPCR Reagents
• Absolute Q dPCR Master Mix (5X), 360 μL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• E.coli DNA Assay Mix (10X), 60 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

Detection Method Primer-probe
Format Kit
PCR Method Digital PCR
Host Cell E.coli
Label or Dye FAM + VIC
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
For Use With (Application) Digital PCR (dPCR)
No. of Reactions 100 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.