missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ dPCR CHO DNA Kit
GREENER_CHOICE

Catalog No. A59366
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A59366 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A59366 Supplier Applied Biosystems™ Supplier No. A59366

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems resDNASEQ dPCR CHO DNA Kit is a powerful digital PCR (dPCR) solution for quantitating residual DNA from CHO expression systems, which are commonly utilized for the production of monoclonal antibodies (mAbs).

The Applied Biosystems resDNASEQ dPCR CHO DNA Kit is a powerful digital PCR (dPCR) solution for quantitating residual DNA from CHO expression systems, which are commonly utilized for the production of monoclonal antibodies (mAbs). By harnessing the capabilities of high-performance TaqMan chemistry, the resDNASEQ CHO assay provides a rapid and reliable approach for accurately quantifying residual CHO DNA. This assay offers exceptional sensitivity and specificity, helping ensure that the quantitation data obtained from a wide range of sample types, including in-process samples with varying sample matrices and final product formulations, is reliable and consistent, instilling confidence in the results.

Features of the resDNASEQ dPCR CHO DNA Kit include:

  • Accurate, absolute quantitation of residual CHO DNA
  • Easy-to-use, integrated sample-to-results system features TaqMan dPCR master mix and TaqMan primer/probe set
  • Highly sensitive quantitation delivers results in about three hours
  • Optional manual or automated sample preparation, optimized for quantitative recovery of samples from complex matrices

Digital PCR workflow
The resDNASEQ dPCR CHO DNA Kit is part of an integrated dPCR workflow for impurity testing during biopharmaceutical manufacturing. Optional use of the Pharma KingFisher Apex 96 Deep-Well Magnetic Particle Processor with the PrepSEQ Nucleic Acid Sample Preparation Kit helps ensure high recovery of residual DNA with less labor and error than manual processing. The Pharma KingFisher Apex instrument can process up to 24 samples in triplicate, compared to three samples in triplicate using a manual method.

To help ensure performance that meets or exceeds regulatory compliance, the resDNASEQ dPCR kit has been internally validated on the QuantStudio Absolute Q Digital PCR System. Data analysis is streamlined using the QuantStudio Absolute Q dPCR Software, which includes accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance.

Specifications

Content And Storage • 5X Absolute Q Digital PCR Master Mix, 0.36 mL; Store at -15°C to -25°C, protect from light
• dPCR CHO DNA Assay Mix, 90 μL; Store at -15°C to -25°C, protect from light
• Negative Control (water), 1 mL; Store at -15°C to -25°C before first use and 2°C to 8°C after first use
Detection Method Digital PCR
Form Frozen
PCR Method Digital PCR
Host Cell CHO
Label or Dye FAM + VIC
Product Line resDNASEQ
Product Type dPCR CHO DNA Kit
Quantity 100 reactions
For Use With (Application) DNA Quantitation

For Research Use Only.