missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resazurin sodium salt

As indicator pH 3.8 (orange) to 6.5 (dark violet) | CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol

$77.82 - $177.58

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym 7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAB2118703
View Documents Promotion Details
Thermo Scientific Chemicals
B2118703
1 g
Each for $77.82
Only null left
 
AAB2118706
View Documents Promotion Details
Thermo Scientific Chemicals
B2118706
5 g
Each for $177.58
Only null left
 
Description

Description

As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

Solubility
Soluble in water, dimethylsulfoxide and ammonium hydroxide.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Specifications

Chemical Identifiers

62758-13-8
251.173
IVGPGQSSDLDOLH-UHFFFAOYSA-M
112939
C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]
C12H6NNaO4
MFCD00005036
7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate

Specifications

62758-13-8
MFCD00005036
14,8141
Soluble in water,dimethylsulfoxide and ammonium hydroxide.
C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]
251.173
251.18
C12H6NNaO4
3794971
7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
IVGPGQSSDLDOLH-UHFFFAOYSA-M
sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
112939
Resazurin sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 263-718-5

RTECSNumber : SP7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only