missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resazurin sodium salt

Catalog No. AAB2118703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2118703 1 g
AAB2118706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2118703 Supplier Thermo Scientific Chemicals Supplier No. B2118703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

As indicator pH 3.8 (orange) to 6.5 (dark violet) | CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol

As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-red

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym 7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Beilstein 3794971
Merck Index 14,8141
Solubility Information Soluble in water,dimethylsulfoxide and ammonium hydroxide.
Quantity 1 g
Formula Weight 251.18
Chemical Name or Material Resazurin sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 263-718-5
RTECSNumber SP7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only