accessibility menu, dialog, popup

UserName

CAS RN 62758-13-8

IUPAC Name: sodium 10-oxo-10H-10λ⁵-phenoxazin-10-ylium-3,7-bis(olate)
Synonyms: sodium 10-oxo-10H-10λ⁵-phenoxazin-10-ylium-3,7-bis(olate)
Molecular Weight (g/mol): 251.17
Molecular Formula: C12H6NNaO4
InChi Key: OPVDVVJXMPZWLP-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=C2C(OC3=CC([O-])=CC=C3[N+]2=O)=C1
Quantity 
  • (4)
  • (2)
  • (4)
Color 
  • (6)
Grade 
  • (6)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (6)
  • (6)
  • (2)
  • (2)