Learn More
Thermo Scientific™ Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
$314.49 - $712.52
Specifications
Content And Storage 2°C to 8°C
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R30854401
View Documents Promotion Details
Thermo Scientific™
RE30854401
100 Tests
Pack of 100 Each for $712.52
Only null left
R30854301
View Documents Promotion Details
Thermo Scientific™
R30854301
50 Test
Pack of 50 Each for $314.49
Only null left
22674073
View Documents Promotion Details
Thermo Scientific™
RE211172
25 Tests/Kit
Pack of 25 Each for $356.49
Only null left
Description
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Specifications
2°C to 8°C