Learn More
Thermo Scientific™ Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Supplier:  Thermo Scientific™ R30854301
 View more versions of this product
Catalog No. R30854301
Only null left
Description
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Chemical Identifiers
2°C to 8°C
Specifications
2°C to 8°C
Yes
50 Test
Product Suggestions
Product Suggestions