Learn More
Thermo Scientific™ Remel™ PYR Disc with Reagent
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Quantity | 25 Tests/Kit |