missing translation for 'onlineSavingsMsg'
Learn More

Regular Unleaded Gasoline, High Level, SPEX CertiPrep™

Catalog No. SRG20K
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SRG20K 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SRG20K Supplier SPEX CertiPrep Group LLC Supplier No. SRG20K

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Regular Unleaded Gas-87octane86290-81-520,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Regular Unleaded Gasoline (High Level)
Packaging 2mL amber prescored glass ampule
CAS 86290-81-5, 67-56-1
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 2.0, 98.0
CAS Max % 2.0, 98.0
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Industry Type Petrochemical Testing
Quantity 1 mL
Show More Show Less

Product intended for laboratory use only