missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 0.5 L

Catalog No. 10569125
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
300 kDa
5 kDa
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
10569125 5 kDa
10569127 2 kDa
10569123 10 kDa
10569121 30 kDa
10569119 100 kDa
10569117 300 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10569125 Supplier Sartorius Supplier No. VFS050H0005IV

Plug-and-play tangential flow filtration cassette that is made from regenerated cellulose, for ultrafiltration and diafiltration of intermediate sample volumes.

  • Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
  • All-in-one design avoids the need for expensive cassette holders and TFF systems.
  • Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
  • High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
  • Improve safety for you and your sample with no need for flushing or cleaning.
  • Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
  • Membrane material: Regenerated cellulose
  • Recommended feed volume: 100 to 500 mL

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Polycarbonate
Material (Membrane) Regenerated Cellulose
MWCO 5 kDa
Sterility Non-sterile
Type TFF cassette
Volume (Metric) 0.5 L
For Use With (Equipment) Peristaltic Pump
Sample Volume 0.1 to 0.5 L