missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Reflective Sheeting Sign Kit: Bradylite NFPA Diamonds

Improve workplace safety with proper signage and markings. This Brady™ reflective sheeting sign kit is a durable, clear and effective tool to raise awareness to workplace hazards.

$109.79 - $280.92

Specifications

Material Reflective Sheeting
Shape Square
Displayed Text Reflective Sheeting Sign Kit: Bradylite NFPA Diamonds
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19110395
View Documents Promotion Details
Brady™
58274
10 x 10 in.
Pack of 10 Each for $280.92
Only null left
 
19110394
View Documents Promotion Details
Brady™
58272
5 x 5 in.
Pack of 10 Each for $109.79
Only null left
 
Description

Description

Specifications

Specifications

Reflective Sheeting
Square
Reflective Sheeting Sign Kit: Bradylite NFPA Diamonds
Videos
SDS
Documents

Documents