missing translation for 'onlineSavingsMsg'
Learn More

Reference Material of Sulfur in Fuel Oil Certified by The Japan Petroleum Institute, TCI America™

Catalog No. S0316100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0316100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0316100ML Supplier TCI America Supplier No. S0316100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfur Content 0mass% level

Chemical Identifiers

MDL Number MFCD00197838

Specifications

UN Number 1268
Quantity 100 mL
Chemical Name or Material Reference Material of Sulfur in Fuel Oil Certified by The Japan Petroleum Institute, Sulfur Content
EINECSNumber (9)-1700
TSCA No