missing translation for 'onlineSavingsMsg'
Learn More

Reference Material of Nitrogen in Residual Fuel Oil Certified by The Japan Petroleum Institute, TCI America™

Catalog No. S025020ML
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S025020ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S025020ML Supplier TCI America Supplier No. S025020ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitrogen Content 0.05mass% level

Chemical Identifiers

MDL Number MFCD00197837

Specifications

Quantity 20 mL
Chemical Name or Material Reference Material of Nitrogen in Residual Fuel Oil Certified by The Japan Petroleum Institute, Nitr