missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.5 N Aqueous Solution, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Hydroxide, 0.5 N Aqueous Solution - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications
Specifications
| CAS Max % | 0.02, 0.98 |
| Concentration | 0.5 N ± 0.005 N |
| Quantity | 500 mL |
| Grade | Chemical Solution |
| Packaging | Plastic |
| Chemical Name or Material | Sodium Hydroxide Solution |