missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 N Solution, APHA, Spectrum™ Chemical

Catalog No. 18606531
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
6 x 1 L
12 x 1 L
200 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18606531 200 L
18606529 500 mL
18606525 4 L
18606530 6 x 1 L
18606526 12 x 1 L
18606527 4 x 4 L
18606528 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18606531 Supplier Spectrum Chemical Mfg Cor Supplier No. S340200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S-340, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 0.100 N Solution, APHA is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.004, 0.996
Concentration 0.1 N ± 0.003 N
Quantity 200 L
Grade APHA
Packaging Plastic
Chemical Name or Material Sodium Hydroxide Solution