missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Active PKA C alpha Protein

Catalog No. 4268KS010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4268KS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4268KS010 Supplier R&D Systems™ Supplier No. 4268KS010

missing translation for 'validMainframeMsg'

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The activity of PKA C alpha is typically 1626-2201 nmol/min/mg using a synthetic peptide substrate.

Specifications

Components Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol.
Purity >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
For Use With (Application) Bioactivity
Storage Buffer Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 10 mM Glutathione, 0.25 mM DTT, 0.1 mM EDTA, 0.1 mM PMSF, and 25% Glycerol.
Content And Storage This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Accession Number NM_002730
Gene ID (Entrez) 5566
Gene Symbol PRKACA
Gene Alias cAMP-dependent protein kinase catalytic subunit alpha, Cs, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, Pk. C-alpha, PKACA, PKCA1, PKCD, PRKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha
Source Spodoptera frugiperda, Sf 9 (baculovirus)-derived human Pk. C alpha protein
Quantity 10 μg
Product Type Recombinant Human cAMP-dependent Protein Kinase Catalytic Subunit alpha
Molecular Weight (g/mol) M.W. (Observed): 69 kDa
Product Line R&D Systems
Show More Show Less