missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), Reagents

Catalog No. p-200021209
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Glass Bottle
Poly Bottle
Poly Drum
Tin Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338147 1 L Poly Bottle
01338146 200 L Poly Drum
01338145 4 L Poly Bottle
01338144 4 L Glass Bottle
01338143 4 L Tin Can
01338142 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01338147 Supplier Labchem Inc Supplier No. C11207001A
Only null left

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -80°C
Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.785
Boiling Point 72°C
Percent Purity Min. 99.5%
Grade ACS
Chemical Name or Material Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol)