missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), Reagents

Catalog No. 01338147
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Glass Bottle
Poly Bottle
Poly Drum
Tin Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338147 1 L Poly Bottle
01338146 200 L Poly Drum
01338145 4 L Poly Bottle
01338144 4 L Glass Bottle
01338143 4 L Tin Can
01338142 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01338147 Supplier Labchem Inc Supplier No. C11207001A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -80°C
Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.785
Boiling Point 72°C
Percent Purity Min. 99.5%
Grade ACS
Chemical Name or Material Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol)