missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), Reagents
Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1120700200E
Specifications
| Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol) | |
| 64-17-5,67-63-0,67-56-1 | |
| 72°C | |
| Poly Drum | |
| 0.785 | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol | |
| Min. 99.5% |
| -80°C | |
| Colorless | |
| 200 L | |
| Liquid | |
| C2H6O | |
| CCO | |
| 46.07 | |
| ACS |
Chemical Identifiers
| 64-17-5 | |
| 46.07 | |
| ethanol |
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| CCO |